H-Glu-Ala-Leu-Phe-Gln-pNA Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4028277.0005
4028277.0025
L-2050.0005BAEA
570.65
CAD
L-2050.0005BA
L-2050.0025BA
H-Glu-Ala-Leu-Phe-Gln-pNA Trifluoroacetate
Proteins and Peptides
The substrate EALFQ-pNA representing the 2C/3A cleavage site of human rhinovirus-14 (HRV-14) polyprotein was hydrolyzed by human rhinovirus-14 3C protease, which represents an attractive target for antiviral drug development. It exhibited a kcat/Km value of 840 M⁻¹s⁻¹.