Ac-YVAD-pNA

Supplier: BACHEM AMERICAS INC

4026201.0005 4026201.0025
L-1890.0005BAEA 413.58 CAD
L-1890.0005BA L-1890.0025BA
Ac-YVAD-pNA
Ac-YVAD-pNA

Sequence: Ac-Tyr-Val-Ala-Asp-pNA
Synonym(s): Ac-YVAD-pNA


Ac-YVAD-pNA, chromogenic substrate for caspase-1 (ICE).

Formula: C₂₉H₃₆N₆O₁₀
Storage Temperature: Freezer
MDL Number: MFCD00274387
CAS Number: 149231-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR