Val-Leu-Arg-p-nitroanilide

Supplier: BACHEM AMERICAS INC

4026051.0050 4026051.0250
L-1885.0050BAEA 793.9 CAD
L-1885.0050BA L-1885.0250BA
Val-Leu-Arg-p-nitroanilide
Val-Leu-Arg-p-nitroanilide

Sequence: H-D-Val-Leu-Arg-pNA · 2 AcOH
Synonym(s): DVal-Leu-Arg-pNA


See also I-1160, I-1960, L-1545, M-1130, and M-1140.


D-VLR-pNA, chromogenic substrate for a convenient, sensitive, and selective assay of glandular kallikrein activity, e.g. of human and rat urinary kallikreins.

Formula: C₂₃H₃₈N₈O₅
Storage Temperature: Freezer
MDL Number: MFCD09250882
CAS Number: 64816-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR