H-Gly-Pro-pNA · HCl
Supplier: BACHEM AMERICAS INC
4025614.0250
4025614.1000
L-1880.0250BAEA
926.18
CAD
L-1880.0250BA
L-1880.1000BA
H-Gly-Pro-pNA · HCl
Proteins and Peptides
See also H-Lys-Pro-AMC (I-1745) and H-Lys-Pro-MNA (J-1255) .
Excellent substrate for dipeptidylpeptidase IV (DPP IV).