L-Valine-p-nitroanilide hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Val-pNA·HCl

4019078.0250 4019078.1000
L-1855.0250BAEA 185.3 CAD
L-1855.0250BA L-1855.1000BA
L-Valine-p-nitroanilide hydrochloride
L-Valine-p-nitroanilide hydrochloride

Sequence: H-Val-pNA · HCl


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.
See also Z-Val-ONp, C-2830.

Formula: C₁₁H₁₆ClN₃O₃
MW: 273.717 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058470
CAS Number: 77835-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR