H-Phe-Pro-Ala-pNA

Supplier: BACHEM AMERICAS INC

Synonyms: H-Phe-Pro-Ala-pNA

4017802.0050 4017802.0250
L-1800.0050BAEA 570.46 CAD
L-1800.0050BA L-1800.0250BA
H-Phe-Pro-Ala-pNA
H-Phe-Pro-Ala-pNA

Sequence: H-Phe-Pro-Ala-pNA


FPA-pNA is cleaved by the sedolisins B, C, and D, tripeptidyl peptidases from Aspergillus fumigatus.

Formula: C₂₃H₂₇N₅O₅
MW: 453.499 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00238130
CAS Number: 201738-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR