H-Ala-Phe-Pro-pNA
Supplier: BACHEM AMERICAS INC
4017695.0050 4017695.0250
L-1795.0050BAEA 439.41 CAD
L-1795.0050BA L-1795.0250BA
H-Ala-Phe-Pro-pNA
Proteins and Peptides
AFP-pNA, substrate for prolyl tripeptidyl aminopeptidases from the periodontal pathogens Porphyromonas gingivalis and Prevotella nigrescens.