Suc-Ala-Ala-Pro-Arg-Pna

Supplier: BACHEM AMERICAS INC

4017320.0050 4017320.0250
L-1720.0050BAEA 1345.09 CAD
L-1720.0050BA L-1720.0250BA
Suc-Ala-Ala-Pro-Arg-Pna
Suc-Ala-Ala-Pro-Arg-Pna

Sequence: Suc-Ala-Ala-Pro-Arg-pNA


Substrates often used for characterizing newly isolated PPIases, subtilisins or other enzymes. Bachem offers Suc-Ala-Ala-Pro-Xaa (Suc-AAPX) substrates containing Ala, Leu, Met, Nle, and Nva (the pancreatic elastase substrates L-1775, L-1390, L-1395, and L-1780), Arg, Asp, Glu, Ile, Lys, Orn, Phe (the PPIase substrates I-1465, L-1400, and M-2305), and Val (the neutrophil elastase substrates I-1490 and L-1770) as Xaa.


Suc-AAPR-pNA, a substrate for wild-type and K188D/D189K trypsins (Km = 32.8 mM respectively 50.8 mM).

Formula: C₂₇H₃₉N₉O₉
Storage Temperature: Freezer
MDL Number: MFCD00238354
CAS Number: 131068-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR