Suc-Phe-Ala-Ala-Phe-pNA
Supplier: BACHEM AMERICAS INC
4013859.0050
4013859.0250
L-1675.0050BAEA
258.14
CAD
L-1675.0050BA
L-1675.0250BA
Suc-Phe-Ala-Ala-Phe-pNA
Proteins and Peptides
This enzyme substrate was used to test the concept of substrate-assisted catalysis with an engineered bacterial serine endopeptidase, subtilisin. Suc-FAAF-pNA has been employed for assaying subtilisins from psychrophilic bacteria and myroilysin, a metalloproteinase from the deep sea bacterium Myroides profundi.