Ac-Ala-pNA
Supplier: BACHEM AMERICAS INC
4014152.0250
4014152.1000
L-1640.0250BAEA
810.05
CAD
L-1640.0250BA
L-1640.1000BA
Ac-Ala-pNA
Ac-Ala-pNA
Sequence: Ac-Ala-pNA
Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).
See also Z-Ala-ONp, C-1040.
AANA, substrate for acylaminoacyl peptidases (APH, acyl peptide hydrolase), an enzyme able to degrade monomeric and oligomeric amyloid peptides
Formula:
C₁₁H₁₃N₃O₄ MW: 251.242 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00057754 CAS Number: 35978-75-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...