Suc-Ala-Glu-Pro-Phe-Pna

Supplier: BACHEM AMERICAS INC

4015607.0050 4015607.0250
L-1635.0050BAEA 1165.06 CAD
L-1635.0050BA L-1635.0250BA
Suc-Ala-Glu-Pro-Phe-Pna
Suc-Ala-Glu-Pro-Phe-Pna

Sequence: Suc-Ala-Glu-Pro-Phe-pNA


Suc-AEPF-pNA, chromogenic substrate for the peptidylprolyl isomerase Pin1.

Formula: C₃₂H₃₈N₆O₁₁
Storage Temperature: Freezer
MDL Number: MFCD00238367
CAS Number: 128802-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR