Suc-Ala-Leu-Pro-Phe-pNA
Supplier: BACHEM AMERICAS INC
4015254.0050
4015254.0250
L-1620.0050BAEA
578.33
CAD
L-1620.0050BA
L-1620.0250BA
Suc-Ala-Leu-Pro-Phe-pNA
Proteins and Peptides
Suc-ALPF-pNA, substrate for FK-506 binding proteins (FKBPs, also called macrophilins) and cyclophilins, which belong to the group of peptidyl prolyl cis-trans isomerases (PPIases). This tetrapeptide has been used for an uncoupled protease-free assay of PPIase activity.