Suc-apppa

Supplier: BACHEM AMERICAS INC

Synonyms: Suc-Ala-Phe-Pro-Phe-pNA

4016001.0050 4016001.0250
L-1605.0050BAEA 903.62 CAD
L-1605.0050BA L-1605.0250BA
Suc-apppa
Suc-apppa

Sequence: Suc-Ala-Phe-Pro-Phe-pNA


Substrate for FK-506 binding proteins (FKBPs, also called macrophilins) and cyclophilins, which belong to the group of peptidyl prolyl cis-trans isomerases (PPIases). This tetrapeptide has been used for an uncoupled protease-free assay of PPIase activity.

Formula: C₃₆H₄₀N₆O₉
MW: 700.749 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00238374
CAS Number: 128802-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR