Methoxycarbonyl-D-Nle-Gly-Arg-Pna

Supplier: BACHEM AMERICAS INC

4012563.0050 4012563.0250
L-1565.0050BAEA 774.5 CAD
L-1565.0050BA L-1565.0250BA
Methoxycarbonyl-D-Nle-Gly-Arg-Pna
Methoxycarbonyl-D-Nle-Gly-Arg-Pna

Sequence: Methoxycarbonyl-D-Nle-Gly-Arg-pNA
Synonym(s): Moc-D-Nle-Gly-Arg-pNA


Substrate for coagulation factor Xa and, in the presence of 33% ethylenene glycol, IXa (Km = 3.62 mM, pH 7.4).

Formula: C₂₂H₃₄N₈O₇
Storage Temperature: Freezer
MDL Number: MFCD02094615
CAS Number: 104186-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR