Z-Gly-Pro-Arg-pNA Acetate, Bachem
Supplier: BACHEM AMERICAS INC
4000768.0025 4000768.0100
L-1560.0025BAEA 570.65 CAD
L-1560.0025BA L-1560.0100BA
Z-Gly-Pro-Arg-pNA Acetate, Bachem
Proteins and Peptides
Z-GPR-pNA, a chromogenic serine protease substrate.