Z-Val-Gly-Arg-pNA Acetate, Bachem
Supplier: BACHEM AMERICAS INC
4007266.0005 4007266.0025
L-1555.0005BAEA 206.51 CAD
L-1555.0005BA L-1555.0025BA
Z-Val-Gly-Arg-pNA Acetate, Bachem
Proteins and Peptides
Z-VGR-pNA, a chromogenic substrate for envelysin and t-plasminogen activator.