H-Glu-Pna

Supplier: BACHEM AMERICAS INC

4008041.0250 4008041.1000
L-1540.0250BAEA 284.04 CAD
L-1540.0250BA L-1540.1000BA
H-Glu-Pna
H-Glu-Pna

Sequence: H-Glu-pNA


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
See also the glutamic acid γ-diazomethyl ketones L-DON (F-1510) and D-DON (F-2185).


Chromogenic substrate of glutamyl aminopeptidase (aminopeptidase A, angiotensinase A).

Formula: C₁₁H₁₃N₃O₅
Storage Temperature: Freezer
MDL Number: MFCD00038111
CAS Number: 24032-35-7
EINECS: 245-994-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR