TOS-Gly-Pro-Lys-PNA
Supplier: BACHEM AMERICAS INC
4010701.0025
4010701.0100
L-1515.0025BAEA
748.67
CAD
L-1515.0025BA
L-1515.0100BA
TOS-Gly-Pro-Lys-PNA
TOS-Gly-Pro-Lys-PNA
Sequence: Tos-Gly-Pro-Lys-pNA
Synonym(s): Tos-GPK-pNA
Chromogenic serine protease substrate, e.g. for tryptase.
Formula:
C₂₆H₃₄N₆O₇S Storage Temperature: Freezer |
MDL Number:
MFCD00057232 CAS Number: 73392-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...