D-Phe-Pip-Arg-PNA

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Phe-Homopro-Arg-pNA·2 acetate

4008145.0050 4008145.0250
L-1490.0050BAEA 877.5 CAD
L-1490.0050BA L-1490.0250BA
D-Phe-Pip-Arg-PNA
D-Phe-Pip-Arg-PNA

Sequence: H-D-Phe-Homopro-Arg-pNA · 2 acetate
Synonym(s): H-D-Phe-Pip-Arg-pNA · 2 acetate#H-D-Phe-L-pipecolyl-Arg-pNA · 2 acetate


Thrombin substrates are often cleaved as well by trypsin and trypsin-like enzymes, e.g. I-1365, I-1560, J-1120, and L-1150.


Chromogenic substrate specific for thrombin but insensitive to factor Xa.

Formula: C₂₇H₃₆N₈O₅
MW: 552.636 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02259651
CAS Number: 115388-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR