Bz-Ile-glu-gly-arg-pna

Supplier: BACHEM AMERICAS INC

4003874.0001 4003874.0005
L-1475.0001BAEA 232.35 CAD
L-1475.0001BA L-1475.0005BA
Bz-Ile-glu-gly-arg-pna
Bz-Ile-glu-gly-arg-pna

Sequence: Bz-Ile-Glu-Gly-Arg-pNA


See also I-1160, I-1960, L-1545, M-1130, and M-1140.


Bz-IEGR-pNA, substrate for plasma kallikrein and coagulation factor XIIa.

Formula: C₃₂H₄₃N₉O₉
Storage Temperature: Freezer
MDL Number: MFCD00133580
CAS Number: 59068-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR