L-Valine-4-nitroanilide

Supplier: BACHEM AMERICAS INC

Synonyms: H-Val-pNA

4004208.0250 4004208.1000
L-1440.0250BAEA 180.91 CAD
L-1440.0250BA L-1440.1000BA
L-Valine-4-nitroanilide
L-Valine-4-nitroanilide

Sequence: H-Val-pNA


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.
See also Z-Val-ONp, C-2830.

Formula: C₁₁H₁₅N₃O₃
MW: 237.259 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00080972
CAS Number: 52084-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR