N-Succinyl-Gly-Gly-Phe-p-nitroanilide

Supplier: BACHEM AMERICAS INC

4002298.0050 4002298.0250
L-1415.0050BAEA 407.1 CAD
L-1415.0050BA L-1415.0250BA
N-Succinyl-Gly-Gly-Phe-p-nitroanilide
N-Succinyl-Gly-Gly-Phe-p-nitroanilide

Sequence: Suc-Gly-Gly-Phe-pNA


Suc-GGF-pNA was used to determine the activity of chymotrypsin and of Streptomyces griseus protease B.

Formula: C₂₃H₂₅N₅O₈
Storage Temperature: Freezer
MDL Number: MFCD00038721
CAS Number: 68982-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR