N-Succinyl-Ala-Ala-Val-Ala p-nitroanilide

Supplier: BACHEM AMERICAS INC

Synonyms: Suc-Ala-Ala-Val-Ala-pNA

4006610.0050 4006610.0250
L-1410.0050BAEA 258.14 CAD
L-1410.0050BA L-1410.0250BA
N-Succinyl-Ala-Ala-Val-Ala p-nitroanilide
N-Succinyl-Ala-Ala-Val-Ala p-nitroanilide

Sequence: Suc-Ala-Ala-Val-Ala-pNA


Suc-AAVA-pNA has been used for assaying hamster chymase 2.

Formula: C₂₄H₃₄N₆O₉
MW: 550.569 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00133618
CAS Number: 108322-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR