Suc-Ala-Ala-Pro-Phe-pNA
Supplier: BACHEM AMERICAS INC
4002299.0050
4002299.0250
4002299.1000
L-1400.0050BAEA
264.44
CAD
L-1400.0050BA
L-1400.0250BA
L-1400.1000BA
Suc-Ala-Ala-Pro-Phe-pNA
Proteins and Peptides
A readily soluble, specific and sensitive substrate for chymotrypsin and human pancreatic elastase. It is also hydrolyzed by cathepsin G and chymase. Furthermore it is the standard substrate for FK-506 binding proteins (FKBPs, also called macrophilins) and cyclophilins, which belong to the group of peptidyl prolyl cis-trans isomerases (PPIases). Thus, Suc-AAPF-pNA has been used for an uncoupled protease-free assay of PPIase activity.