Suc-Ala-Ala-Pro-Met-pNA
Supplier: BACHEM AMERICAS INC
4006760.0050
4006760.0250
L-1395.0050BAEA
645.43
CAD
L-1395.0050BA
L-1395.0250BA
Suc-Ala-Ala-Pro-Met-pNA
Proteins and Peptides
See also I-1270, L-1000, L-1090, L-1180, L-1335, L-1385, L-1400, M-1170, and M-1250.
Suc-AAPM-pNA, a sensitive substrate for human pancreatic elastase.