Suc-Ala-Ala-Pro-Leu-Pna

Supplier: BACHEM AMERICAS INC

4003646.0050 4003646.0250
L-1390.0050BAEA 264.68 CAD
L-1390.0050BA L-1390.0250BA
Suc-Ala-Ala-Pro-Leu-Pna
Suc-Ala-Ala-Pro-Leu-Pna

Sequence: Suc-Ala-Ala-Pro-Leu-pNA


See also I-1270, L-1000, L-1090, L-1180, L-1335, L-1385, L-1400, M-1170, and M-1250.


Suc-AAPL-pNA, a sensitive substrate for human and especially porcine pancreatic elastases. Substrate for leucine endopeptidase from the psychrotropic bacillus B. cereus.

Formula: C₂₇H₃₈N₆O₉
MW: 590.634 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077159
CAS Number: 70968-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR