Suc-Ala-Ala-Ala-Pna
Supplier: BACHEM AMERICAS INC
Synonyms:
4-(((S)-1-(((S)-1-(((S)-1-((4-Nitrophenyl)amino)-1-oxopropan-2-yl)amino)-1-oxopropan-2-yl)amino)-1-oxopropan-2-yl)amino)-4-oxobutanoic acid, Suc-AAA-pNA
Sequence: Suc-Ala-Ala-Ala-pNA
See also C-1040, C-2345, I-1000, and L-1435.
Suc-AAA-pNA, a readily soluble and sensitive substrate for human and rat leukocyte and porcine pancreatic elastases. The trialanine substrate is also hydrolyzed by proteinase K, subtilisins and thermitase as well as by astacin, a crayfish zinc-endopeptidase.
Formula:
C₁₉H₂₅N₅O₈ MW: 451.436 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00036774 CAS Number: 52299-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...