Pyr-pNA
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-N-(4-Nitrophenyl)-5-oxopyrrolidine-2-carboxamide
4002484.0250
4002484.1000
L-1375.0250BAEA
309.81
CAD
L-1375.0250BA
L-1375.1000BA
Pyr-pNA
Pyr-pNA
Sequence: Pyr-pNA
Also called 5-oxoproline or pidolic acid. Bachem uses the acronym Pyr for denoting pyroglutamic acid. Further abbreviations as Glp, Pgl, pGlu (pE), or < Glu (< E) are also in use.
A chromogenic substrate for pyroglutamyl peptidase I.
Formula:
C₁₁H₁₁N₃O₄ MW: 249.23 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00037884 CAS Number: 66642-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...