N-Methoxysuccinyl-Ala-Ala-Pro-Val-p-nitroanilide

Supplier: BACHEM AMERICAS INC

Synonyms: MeOSuc-Ala-Ala-Pro-Val-pNA

4004221.0050 4004221.0250
L-1335.0050BAEA 661.51 CAD
L-1335.0050BA L-1335.0250BA
N-Methoxysuccinyl-Ala-Ala-Pro-Val-p-nitroanilide
N-Methoxysuccinyl-Ala-Ala-Pro-Val-p-nitroanilide

Sequence: MeOSuc-Ala-Ala-Pro-Val-pNA


An excellent, highly soluble and specific substrate for a routine assay of human and rat leukocyte and porcine pancreatic elastases. MeOSuc-AAPV-pNA is also cleaved by myeloblastin.

Formula: C₂₇H₃₈N₆O₉
MW: 590.634 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077137
CAS Number: 70967-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR