L-Leucine-p-nitroanilide

Supplier: BACHEM AMERICAS INC

4001072.0005 4001072.0025
L-1305.0005BAEA 232.35 CAD
L-1305.0005BA L-1305.0025BA
L-Leucine-p-nitroanilide
L-Leucine-p-nitroanilide

Sequence: H-Leu-pNA


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).
Commonly used protease inhibitors as bestatin and the E-64s are derived from leucine. See also H-Leu-NH₂ (E-2650) and Z-Leu-ONp (C-2120).


Substrate for aminopeptidase M and leucine aminopeptidase.

Formula: C₁₂H₁₇N₃O₃
Storage Temperature: Freezer
MDL Number: MFCD00063681
CAS Number: 4178-93-2
EINECS: 224-047-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR