H-D-Ile-Phe-Lys-pNA Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4008063.0050
4008063.0250
L-1300.0050BAEA
1729.74
CAD
L-1300.0050BA
L-1300.0250BA
H-D-Ile-Phe-Lys-pNA Trifluoroacetate
Proteins and Peptides
Patented by Chromogenix AB, Sweden (CH 622 285). A highly specific substrate for human plasmin. For rat plasmin, Heinemann et al. determined a Km of 18.3 μM. Also cleaved by transmembrane tryptase (TMT).