(S)-N-(2-Aminoacetyl)-1-(4-nitrophenyl)pyrrolidine-2-carboxamide-4-methylbenzenesulfonate
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Gly-Pro-pNA·p-tosylate
4003654.0100
4003654.0250
L-1295.0100BAEA
335.58
CAD
L-1295.0100BA
L-1295.0250BA
(S)-N-(2-Aminoacetyl)-1-(4-nitrophenyl)pyrrolidine-2-carboxamide-4-methylbenzenesulfonate
(S)-N-(2-Aminoacetyl)-1-(4-nitrophenyl)pyrrolidine-2-carboxamide-4-methylbenzenesulfonate
Sequence: H-Gly-Pro-pNA · p-tosylate
See also H-Lys-Pro-AMC (I-1745) and H-Lys-Pro-MNA (J-1255) .
Very good substrate for the assay of dipeptidyl-peptidase IV (DPP IV, X-prolyldipeptidyl aminopeptidase), especially in normal and pathological human sera.
Formula:
C₂₀H₂₄N₄O₇S MW: 464.493 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00039007 CAS Number: 65096-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...