H-Gly-Phe-Pna

Supplier: BACHEM AMERICAS INC

4008150.0100 4008150.0250
L-1290.0100BAEA 387.21 CAD
L-1290.0100BA L-1290.0250BA
H-Gly-Phe-Pna
H-Gly-Phe-Pna

Sequence: H-Gly-Phe-pNA


Substrate for dipeptidyl aminopeptidase I (cathepsin C).

Formula: C₁₇H₁₈N₄O₄
Storage Temperature: Freezer
MDL Number: MFCD00038182
CAS Number: 21027-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR