H-Gly-Arg-pNA · 2 HCl
Supplier: BACHEM AMERICAS INC
4007356.0050 4007356.0250
L-1285.0050BAEA 335.58 CAD
L-1285.0050BA L-1285.0250BA
H-Gly-Arg-pNA · 2 HCl
Proteins and Peptides
GR-pNA, chromgenic substrate for DPP I (cathepsin C) and DPP II, also cleaved by P. gingivalis dipeptidyl peptidase.