Z-Tyr-Lys-Arg-pNA·2 TFA

Supplier: BACHEM AMERICAS INC

4005988.0050 4005988.0250
L-1250.0050BAEA 661.51 CAD
L-1250.0050BA L-1250.0250BA
Z-Tyr-Lys-Arg-pNA·2 TFA
Z-Tyr-Lys-Arg-pNA·2 TFA

Sequence: Z-Tyr-Lys-Arg-pNA · 2 TFA


The chromogenic substrate Z-YKR-pNA is cleaved by proteinase yscF, now known as the Kex2 endoprotease kexin, a membrane-bound yeast enzyme responsible for processing of the α-factor precursor protein.

Formula: C₃₅H₄₅N₉O₈·2C₂HF₃O₂
MW: 947.85 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00238515
CAS Number: 108318-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR