Z-Phe-Arg-pNA · HCl
Supplier: BACHEM AMERICAS INC
4009170.0050
4009170.0250
L-1242.0050BAEA
313.55
CAD
L-1242.0050BA
L-1242.0250BA
Z-Phe-Arg-pNA · HCl
Proteins and Peptides
Z-FR-pNA, a sensitive chromogenic substrate for cathepsin L and papain. Z-FR-pNA is also cleaved by falcipain 2. CAS Number (net): 117761-01-0.