Z-Gly-Pro-PNA

Supplier: BACHEM AMERICAS INC

4003649.0250 4003649.1000
L-1235.0250BAEA 275.69 CAD
L-1235.0250BA L-1235.1000BA
Z-Gly-Pro-PNA
Z-Gly-Pro-PNA

Sequence: Z-Gly-Pro-pNA


See also I-1350 and K-1490.


Chromogenic substrate for post-proline cleaving enzyme (prolyl endopeptidase).

Formula: C₂₁H₂₂N₄O₆
MW: 426.429 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038815
CAS Number: 65022-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR