Z-Arg-pNA·HCl
Supplier: BACHEM AMERICAS INC
Synonyms:
Z-Arg-pNA·HCl
4000780.0001
4000780.0005
L-1220.0001BAEA
309.81
CAD
L-1220.0001BA
L-1220.0005BA
Z-Arg-pNA·HCl
Z-Arg-pNA·HCl
Sequence: Z-Arg-pNA · HCl
Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).
Synonym(s): L-ZAPA
Formula:
C₂₀H₂₅ClN₆O₅ Storage Temperature: Freezer |
MDL Number:
MFCD00058247 CAS Number: 59188-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...