Endotoxin Substrate Acetate
Supplier: BACHEM AMERICAS INC
4007683.0050
4007683.0250
L-1195.0050BAEA
562.76
CAD
L-1195.0050BA
L-1195.0250BA
Endotoxin Substrate Acetate
Proteins and Peptides
Boc-LGR-pNA, a chromogenic substrate for horseshoe crab clotting enzyme, which is used in quantitative assays of endotoxin.