Boc-Ala-Ala-PNA

Supplier: BACHEM AMERICAS INC

4006301.0250 4006301.1000
L-1160.0250BAEA 476.62 CAD
L-1160.0250BA L-1160.1000BA
Boc-Ala-Ala-PNA
Boc-Ala-Ala-PNA

Sequence: Boc-Ala-Ala-pNA


Boc-AA-pNA is cleaved by serine proteases.

Formula: C₁₇H₂₄N₄O₆
MW: 380.401 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038112
CAS Number: 50439-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR