BZ-Tyr-PNA

Supplier: BACHEM AMERICAS INC

4001259.0001 4001259.0005
L-1155.0001BAEA 309.66 CAD
L-1155.0001BA L-1155.0005BA
BZ-Tyr-PNA
BZ-Tyr-PNA

Sequence: Bz-Tyr-pNA


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
See also Boc-Tyr-ONp, M-1180.


Synonym(s): L-BTpNA

Formula: C₂₂H₁₉N₃O₅
Storage Temperature: Freezer
MDL Number: MFCD00038109
CAS Number: 6154-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR