Bz-Phe-Val-Arg-pNA · HCl
Supplier: BACHEM AMERICAS INC
4002044.0050 4002044.0250
L-1150.0050BAEA 529.25 CAD
L-1150.0050BA L-1150.0250BA
Bz-Phe-Val-Arg-pNA · HCl
Proteins and Peptides
Thrombin substrates are often cleaved as well by trypsin and trypsin-like enzymes, e.g. I-1365, I-1560, J-1120, and L-1150.
A very good substrate for thrombin, trypsin, and papain. Bz-FVR-pNA cleavage by thrombin can also be used for a simple photometric assay of antithrombin III (heparin cofactor). Chromogenic substrate for cruzipain and subtilisin.