N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride, BAPNA, BANI, Bz-DL-Arg-pNA·HCl

4001028.0005 4001028.0025
L-1140.0005BAEA 187.69 CAD
L-1140.0005BA L-1140.0025BA
N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride
N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride

Sequence: Bz-DL-Arg-pNA · HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Synonym(s): DL-BAPA · HCl#DL-BApNA · HCl

Formula: C₁₉H₂₃ClN₆O₄
Storage Temperature: Freezer
MDL Number: MFCD00012846
CAS Number: 911-77-3
EINECS: 213-011-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR