BZ-D-Arg-PNAHCl
Supplier: BACHEM AMERICAS INC
Synonyms:
Bz-D-Arg-pNA·HCl
4009629.0025
4009629.0250
L-1135.0025BAEA
397.18
CAD
L-1135.0025BA
L-1135.0250BA
BZ-D-Arg-PNAHCl
BZ-D-Arg-PNAHCl
Sequence: Bz-D-Arg-pNA · HCl
Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).
Synonym(s): D-BAPA · HCl#D-BApNA · HCl
Formula:
C₁₉H₂₃ClN₆O₄ MW: 434.881 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00070232 CAS Number: 21653-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...