N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride, Bz-Arg-pNA·HCl, Bz-Arg-PNA.HCl, N-α-Benzoyl-L-Arginine p-Nitroanilide Hydrochloride, (S)-N-(5-Guanidino-1-((4-nitrophenyl)amino)-1-oxopentan-2-yl)benzamide hydrochloride

4000792.0250 4000792.1000
L-1130.0250BAEA 183.98 CAD
L-1130.0250BA L-1130.1000BA
N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride

Sequence: Bz-Arg-pNA · HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Synonym(s): L-BApNA · HCl#L-BAPA · HCl

Formula: C₁₉H₂₃ClN₆O₄
MW: 434.881 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063682
CAS Number: 21653-40-7
EINECS: 244-505-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR