(S)-2-Amino-5-guanidino-N-(4-nitrophenyl)pentanamide dihydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: L-Arginine 4-nitroanilide dihydrochloride, H-Arg-pNA·2 HCl, L-Arginine p-nitroanilide dihydrochloride, H-Arg-pNA·2HCl

4000788.0250 4000788.1000
L-1120.0250BAEA 180.91 CAD
L-1120.0250BA L-1120.1000BA
(S)-2-Amino-5-guanidino-N-(4-nitrophenyl)pentanamide dihydrochloride
(S)-2-Amino-5-guanidino-N-(4-nitrophenyl)pentanamide dihydrochloride

Sequence: H-Arg-pNA · 2 HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Specific chromogenic substrate for cathepsin H.

Formula: C₁₂H₂₀Cl₂N₆O₃
MW: 367.231 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058249
CAS Number: 40127-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR