H-Ala-Pro-pNA
Supplier: BACHEM AMERICAS INC
4002751.0250 4002751.1000
L-1115.0250BAEA 309.81 CAD
L-1115.0250BA L-1115.1000BA
H-Ala-Pro-pNA
Proteins and Peptides
See also H-Lys-Pro-AMC (I-1745) and H-Lys-Pro-MNA (J-1255) .
Very good substrate for the assay of dipeptidyl aminopeptidase IV (DPP IV). It is also hydrolyzed by the thermosensitive dipeptidyl aminopeptidase yscV.