Ala-Ala-Phe-p-nitroanilide
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Ala-Ala-Phe-pNA
4002753.0050
4002753.0250
L-1095.0050BAEA
180.91
CAD
L-1095.0050BA
L-1095.0250BA
Ala-Ala-Phe-p-nitroanilide
Ala-Ala-Phe-p-nitroanilide
Sequence: H-Ala-Ala-Phe-pNA
Synonym(s): AAF-pNA
Substrate for chymotrypsin and tripeptidyl peptidases I and II.
Formula:
C₂₁H₂₅N₅O₅ MW: 427.461 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00057666 CAS Number: 61043-41-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...