L-Alanine-4-nitroanilide hydrochloride

Supplier: BACHEM AMERICAS INC

4026954.0001 4026954.0005
L-1075.0001BAEA 221.88 CAD
L-1075.0001BA L-1075.0005BA
L-Alanine-4-nitroanilide hydrochloride
L-Alanine-4-nitroanilide hydrochloride

Sequence: H-Ala-pNA · HCl


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).
See also Z-Ala-ONp, C-1040.

Formula: C₉H₁₂ClN₃O₃
Storage Temperature: Freezer
MDL Number: MFCD00039088
CAS Number: 31796-55-1
EINECS: 250-811-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR