Alanine-4-nitroanilide
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Ala-pNA
4000210.0001
4000210.0005
L-1070.0001BAEA
317.52
CAD
L-1070.0001BA
L-1070.0005BA
Alanine-4-nitroanilide
Alanine-4-nitroanilide
Sequence: H-Ala-pNA
Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).
See also Z-Ala-ONp, C-1040.
A good substrate for aminopeptidase M (microsomal alanyl aminopeptidase).
Formula:
C₉H₁₁N₃O₃ MW: 209.205 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00053640 CAS Number: 1668-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...